Suc-LLVY-AMC
Lieferant: AAT BIOQUEST
13453
AATB13453EA 152 EUR
AATB13453
Suc-LLVY-AMC
Proteine und Peptide
Suc-LLVY-AMC is a sentive fluorogenic substrate for 20S proteasome, calpains and other chymotrypsin-like proteases.
  • High fluorescence
  • Rapid activation