(Ac-ANW)2R110
Lieferant: AAT BIOQUEST
|
Achtung
|
13455
AATB13455EA
307
EUR
AATB13455
(Ac-ANW)2R110
(Ac-ANW)2R110
(Ac-ANW)2R110 is a selective fluorogenic substrate for proteasome 20S-beta 5i.
- Highly sensitive substrate
- Bright signal
- Multiple applications
ADR:
5,II |