(Ac-ANW)2R110
Lieferant: AAT BIOQUEST
Achtung
13455
AATB13455EA 307 EUR
AATB13455
(Ac-ANW)2R110
(Ac-ANW)2R110
(Ac-ANW)2R110 is a selective fluorogenic substrate for proteasome 20S-beta 5i.
  • Highly sensitive substrate
  • Bright signal
  • Multiple applications
ADR: 5,II