L-Alanine 7-amido-4-methylcoumarin trifluoroacetate salt

Lieferant: AAT BIOQUEST
Achtung

Synonyme: L-Alanine-AMC.TFA

13458
AATB13458EA 152 EUR
AATB13458
L-Alanine 7-amido-4-methylcoumarin trifluoroacetate salt
L-Alanine 7-amido-4-methylcoumarin trifluoroacetate salt

Ala-AMC is a sensitive fluorogenic protease substrate.


  • Protease substrate
  • Bright signal
  • Multiple applications

Formel: C₁₃H₁₄N₂O₃·CF₃CO₂H
Molecular Weight: 360,285 g/mol
MDL: MFCD00037394
CAS-Nummer: 96594-10-4
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR