MeO-Succ-Arg-Pro-Tyr-AMC
Lieferant: AAT BIOQUEST
Achtung
13460
AATB13460EA 152 EUR
AATB13460
MeO-Succ-Arg-Pro-Tyr-AMC
MeO-Succ-Arg-Pro-Tyr-AMC
MeO-Succ-Arg-Pro-Tyr-AMC is a sensitive fluorogenic substrate for chymotrypsin-like proteases.
  • Protease substrate
  • Bright signal
  • Multiple applications
ADR: 5,II