MeO-Succ-Arg-Pro-Tyr-AMC
Lieferant: AAT BIOQUEST
|
Achtung
|
13460
AATB13460EA
152
EUR
AATB13460
MeO-Succ-Arg-Pro-Tyr-AMC
MeO-Succ-Arg-Pro-Tyr-AMC
MeO-Succ-Arg-Pro-Tyr-AMC is a sensitive fluorogenic substrate for chymotrypsin-like proteases.
- Protease substrate
- Bright signal
- Multiple applications
ADR:
5,II |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)