BOC-Val-Pro-Arg-AMC
Lieferant: AAT BIOQUEST
Achtung
13461
AATB13461EA 152 EUR
AATB13461
BOC-Val-Pro-Arg-AMC
BOC-Val-Pro-Arg-AMC
BOC-Val-Pro-Arg-AMC is a sensitive fluorogenic substrate for alpha-thrombin.
  • Alpha-thrombin substrate
  • Bright signal
  • Multiple applications
ADR: 5,II