(BOC-VPR)2R110
Lieferant: AAT BIOQUEST
Achtung
13462
AATB13462EA 152 EUR
AATB13462
(BOC-VPR)2R110
(BOC-VPR)2R110
(BOC-VPR)2R110 is a sensitive fluorogenic substrate for alpha-thrombin.
  • Alpha-thrombin substrate
  • Bright signal
  • Multiple applications
ADR: 5,II