(BOC-VPR)2R110
Lieferant: AAT BIOQUEST
|
Achtung
|
13462
AATB13462EA
152
EUR
AATB13462
(BOC-VPR)2R110
(BOC-VPR)2R110
(BOC-VPR)2R110 is a sensitive fluorogenic substrate for alpha-thrombin.
- Alpha-thrombin substrate
- Bright signal
- Multiple applications
ADR:
5,II |