(Z-LLE)2R110
Lieferant: AAT BIOQUEST
|
Achtung
|
13466
AATB13466EA
152
EUR
AATB13466
(Z-LLE)2R110
(Z-LLE)2R110
The non-fluorescent R110 substrates generate the bright green fluorescent rhodamine 110 product that has Ex/Em = 494/521 nm, and can be easily detected with a FITC filter set.
- Protease substrate
- Bright signal
- Multiple applications
ADR:
5,II |