(Ac-WLA)2R110
Lieferant: AAT BIOQUEST
Achtung
13468
AATB13468EA 152 EUR
AATB13468
(Ac-WLA)2R110
(Ac-WLA)2R110
The non-fluorescent R110 substrates generate the bright green fluorescent rhodamine 110 product that has Ex/Em = 494/521 nm, and can be easily detected with a FITC filter set.
  • Protease substrate
  • Bright signal
  • Multiple applications
ADR: 5,II