TF3-DEVD-FMK
Lieferant: AAT BIOQUEST
Achtung
13474
AATB13474EA 152 EUR
AATB13474
TF3-DEVD-FMK
TF3-DEVD-FMK
TF3-DEVD-FMK is a superior replacement for 5-TAMRA-DEVD-FMK, i.e., 5-carboxytetramethylrhdamine-Asp-Glu-Val-Asp-fluoromethylketone since it can be better excited at 555 nm.
  • Caspase probe
  • Bright signal
  • Multiple applications
ADR: 5,II