TF3-DEVD-FMK
Lieferant: AAT BIOQUEST
|
Achtung
|
13474
AATB13474EA
152
EUR
AATB13474
TF3-DEVD-FMK
TF3-DEVD-FMK
TF3-DEVD-FMK is a superior replacement for 5-TAMRA-DEVD-FMK, i.e., 5-carboxytetramethylrhdamine-Asp-Glu-Val-Asp-fluoromethylketone since it can be better excited at 555 nm.
- Caspase probe
- Bright signal
- Multiple applications
ADR:
5,II |