510-VAD-FMK, mFluor™
Lieferant: AAT BIOQUEST
Achtung
13476
AATB13476EA 152 EUR
AATB13476
510-VAD-FMK, mFluor™
510-VAD-FMK
FAM-VAD is a green fluorescent cell-permeable polycaspase inhibitor to target caspases 1, 2, 3, 6, 8, 9, or 10.
  • Caspase inhibitor
  • Bright signal
  • Multiple applications
ADR: 5,II