510-VAD-FMK, mFluor™
Lieferant: AAT BIOQUEST
|
Achtung
|
13476
AATB13476EA
152
EUR
AATB13476
510-VAD-FMK, mFluor™
510-VAD-FMK
FAM-VAD is a green fluorescent cell-permeable polycaspase inhibitor to target caspases 1, 2, 3, 6, 8, 9, or 10.
- Caspase inhibitor
- Bright signal
- Multiple applications
ADR:
5,II |