Benzoyl-Arg-AMC
Lieferant: AAT BIOQUEST
Achtung
13478
AATB13478EA 152 EUR
AATB13478
Benzoyl-Arg-AMC
Benzoyl-Arg-AMC
This Benzoyl-Arg-AMC is a fluorogenic aminopeptidase and endopeptidase substrate used for the detection of enzymatic activity of human alpha-thrombin.
  • Alpha-thrombin detection
  • Bright signal
  • Multiple applications
ADR: 5,II