Bis-L-Aspartic acid Beta-rhodamine 110
Lieferant: AAT BIOQUEST
|
Achtung
|
13480
AATB13480EA
307
EUR
AATB13480
Bis-L-Aspartic acid Beta-rhodamine 110
Bis-L-Aspartic acid Beta-rhodamine 110
Bis-L-Aspartic acid β-rhodamine 110 is a sensitive fluorogenic substrate for studying lysosomal glycoasparaginases (aspartylglucosaminidases) and L-asparaginases.
- Asparaginase detection
- Bright signal
- Multiple applications
ADR:
5,II |