Bis-L-Aspartic acid Beta-rhodamine 110
Lieferant: AAT BIOQUEST
Achtung
13480
AATB13480EA 307 EUR
AATB13480
Bis-L-Aspartic acid Beta-rhodamine 110
Bis-L-Aspartic acid Beta-rhodamine 110
Bis-L-Aspartic acid β-rhodamine 110 is a sensitive fluorogenic substrate for studying lysosomal glycoasparaginases (aspartylglucosaminidases) and L-asparaginases.
  • Asparaginase detection
  • Bright signal
  • Multiple applications
ADR: 5,II