Bis-Arg-rhodamine 110
Lieferant: AAT BIOQUEST
Achtung
13481
AATB13481EA 460 EUR
AATB13481
Bis-Arg-rhodamine 110
Bis-Arg-rhodamine 110
This Bis Arg-rhodamine 110 is a fluorogenic aminopeptidase and endopeptidase substrate used for the detection of enzymatic activity of human alpha-thrombin.
  • Alpha-thrombin substrate
  • Bright signal
  • Multiple applications
ADR: 5,II