FAM-cGMP PDE Substrat "Green Fluorescence"
Lieferant: AAT Bioquest
|
Achtung
|
13604
AATB13604EA
460
EUR
AATB13604
FAM-cGMP PDE Substrat "Green Fluorescence"
FAM-cGMP PDE
This green fluorescent cGMP derivative is a specific substrate for phosphodiesterase (PDE) V.
- Excellent and specific fluorogenic PDE substrate
- cGMP research tool
- Multiple applications
ADR:
5,II |