FAM-cGMP PDE Substrat "Green Fluorescence"
Lieferant: AAT Bioquest
Achtung
13604
AATB13604EA 460 EUR
AATB13604
FAM-cGMP PDE Substrat "Green Fluorescence"
FAM-cGMP PDE
This green fluorescent cGMP derivative is a specific substrate for phosphodiesterase (PDE) V.
  • Excellent and specific fluorogenic PDE substrate
  • cGMP research tool
  • Multiple applications
ADR: 5,II