GMP-Fluorescein Konjugat zur Kalibrierung
Lieferant: AAT Bioquest
Achtung
13611
AATB13611EA 769 EUR
AATB13611
GMP-Fluorescein Konjugat zur Kalibrierung
GMP-Fluorescein
GMP-Fluorescein conjugate calibrator is desigend to use with green fluorescent FAM-cGMP PDE V substrate, a cGMP derivative that is a specific substrate for phosphodiesterase (PDE) V.
  • Pde V specific
  • High quality
  • Multiple applications
Bitte beachten For research use only.
ADR: 5,II