iFluor® 790 azide
Lieferant: AAT BIOQUEST
1363
AATB1363EA 309 EUR
AATB1363
iFluor® 790 azide
iFluor® 790 azide
iFluor® 790 Azide readily reacts with biomolecules that contain an alkyne group.
  • Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only