iFluor® 790, Maleimide
Lieferant: AAT BIOQUEST
1366
AATB1366EA
460
EUR
AATB1366
iFluor® 790, Maleimide
iFluor®
In vivo fluorescence imaging uses a sensitive camera to detect fluorescence emission from fluorophores in whole-body living small animals.
- Bright Fluorescence
- High Photostability
- Low Quenching On Proteins
- Useful For In-Vivo Imaging
Bitte beachten For research use only.