iFluor® 770 succinimidyl ester
Lieferant: AAT BIOQUEST
1369
AATB1369EA
309
EUR
AATB1369
iFluor® 770 succinimidyl ester
iFluor® 770 succinimidyl ester
iFluor® 770 dyes have fluorescence excitation and emission maxima of ~777 nm and ~797 nm respectively.
- Stable for 12 months
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, particularly antibodies.
Bitte beachten For Research Use Only