iFluor® 770 maleimide
Lieferant: AAT BIOQUEST
1370
AATB1370EA 464 EUR
AATB1370
iFluor® 770 maleimide
iFluor® 770 maleimide
iFluor® 770 dyes have fluorescence excitation and emission maxima of ~777 nm and ~797 nm respectively.
  • Stable for 12 months
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, particularly antibodies.
Bitte beachten For Research Use Only