iFluor® 780 succinimidyl ester
Lieferant: AAT BIOQUEST
1371
AATB1371EA 309 EUR
AATB1371
iFluor® 780 succinimidyl ester
iFluor® 780 succinimidyl ester
iFluor® 780 dyes have fluorescence excitation and emission maxima of ~784 nm and ~808 nm respectively.
  • Stable for 12 months
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, particularly antibodies.
Bitte beachten For Research Use Only
Bestellen
TECHNISCHE DATEN
Environmentally Preferable