iFluor® 780 maleimide
Lieferant: AAT BIOQUEST
1372
AATB1372EA
464
EUR
AATB1372
iFluor® 780 maleimide
iFluor® 780 maleimide
iFluor® 780 dyes have fluorescence excitation and emission maxima of ~784 nm and ~808 nm respectively.
- Stable for 12 months
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, particularly antibodies.
Bitte beachten For Research Use Only