iFluor® 800, Maleimide
Lieferant: AAT BIOQUEST
Achtung
1378
AATB1378EA 460 EUR
AATB1378
iFluor® 800, Maleimide
iFluor®
In vivo fluorescence imaging uses a sensitive camera to detect fluorescence emission from fluorophores in whole-body living small animals.
  • Bright and photostable
  • Minimal quenching
  • Stable conjugation
ADR: 5,II