iFluor® 800, Maleimide
Lieferant: AAT BIOQUEST
|
Achtung
|
1378
AATB1378EA
460
EUR
AATB1378
iFluor® 800, Maleimide
iFluor®
In vivo fluorescence imaging uses a sensitive camera to detect fluorescence emission from fluorophores in whole-body living small animals.
- Bright and photostable
- Minimal quenching
- Stable conjugation
ADR:
5,II |