Amplite® Fluorimetric D-Lactate Dehydrogenase (LDH) Assay Kit
Lieferant: AAT BIOQUEST
13808
AATB13808EA 460 EUR
AATB13808
Amplite® Fluorimetric D-Lactate Dehydrogenase (LDH) Assay Kit
Tests Enzymtests
Lactate dehydrogenase (LDH) is an oxidoreductase enzyme that catalyses the interconversion of pyruvate and lactate.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Bitte beachten For research use only.