Amplite® Colorimetric L-Lactate Dehydrogenase (LDH) Assay Kit
Lieferant: AAT BIOQUEST
13813
AATB13813EA
460
EUR
AATB13813
Amplite® Colorimetric L-Lactate Dehydrogenase (LDH) Assay Kit
Tests
Enzymtests
Lactate dehydrogenase (LDH) is an oxidoreductase enzyme that catalyses the interconversion of pyruvate and lactate.
- In-house QC testing
- Sensitive
- Suggested protocol available
Bitte beachten For research use only.