iFluor® 830 acid
Lieferant: AAT BIOQUEST
1382
AATB1382EA 309 EUR
AATB1382
iFluor® 830 acid
iFluor® 830 acid
iFluor® 830 dye emission is a unique color for spectrum flow cytometry as it is well separated from commonly used far-red fluorophores such as Cy5, Cy7, or allophycocyanin (APC), facilitating multicolor analysis.
  • Stable for 12 months
In vivo fluorescence imaging uses a sensitive camera to detect the fluorescence emission from fluorophores in whole-body living small animals.
Bitte beachten For Research Use Only