Amplite® Fluorimetric Hypochlorite (Hypochlorous Acid) Assay Kit
Lieferant: AAT BIOQUEST
13846
AATB13846EA
460
EUR
AATB13846
Amplite® Fluorimetric Hypochlorite (Hypochlorous Acid) Assay Kit
Tests
Enzymtests
Hypochlorite anion (ClO-) and its protonated form, hypochlorous acid (HClO) are critical reactive oxygen species (ROS) in biological systems.
- In-house QC testing
- Sensitive
- Suggested protocol available
Bitte beachten For research use only.