iFluor® 820, Säure
Lieferant: AAT BIOQUEST
Achtung
1395
AATB1395EA 307 EUR
AATB1395
iFluor® 820, Säure
iFluor®
In vivo fluorescence imaging uses a sensitive camera to detect fluorescence emission from fluorophores in whole-body living small animals.
  • Bright and photostable
  • Minimal quenching
  • Stable conjugation
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=41461dcc41976898a08bc01cdf1db5c61216e18c&vid=4676aaea248fe68d2ed58ddfeb21e85dd74a48af&prid=web&clid=SAPDAM