Fluorescein di-b-D-galactopyranoside (FDG) ≥95%

Lieferant: AAT BIOQUEST
Achtung

14001
AATB14001EA 230 EUR
AATB14001
Fluorescein di-b-D-galactopyranoside (FDG) ≥95%
Fluorescein di-b-D-galactopyranoside (FDG)

Although chromogenic assays of beta-galactosidase activity (i.e. X-Gal) are still widely used, the recent advance in fluorescence instrumentation has made fluorogenic substrates (such as FDG and MUG) be several orders of magnitude more sensitive. FDG is considered to be one of the most sensitive fluorogenic substrates available for detecting beta-galactosidase.

Formel: C₃₂H₃₂O₁₅
CAS-Nummer: 17817-20-8
ADR: 5,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR