4-Methylumbelliferyl alpha-D-galactopyranoside
Lieferant: AAT BIOQUEST
14017
AATB14017EA
133
EUR
AATB14017
4-Methylumbelliferyl alpha-D-galactopyranoside
4-Methylumbelliferyl alpha-D-galactopyranoside
4-Methylumbelliferyl α-D-glucopyranoside is used as a flurogenic substrate for the identification, characterization and kinetic analysis of α-D-glucosidases.
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only