Resorufin-β-D-galactopyranoside ≥95%
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
7-(((2S,3R,4S,5R,6R)-3,4,5-Trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-3H-phenoxazin-3-one, Resorufin β-D-Galactopyranoside
14019
AATB14019EA
152
EUR
AATB14019
Resorufin-β-D-galactopyranoside ≥95%
Resorufin-β-D-galactopyranoside
Resorufin beta-D-galactopyranoside is a fluorogenic galactosidase substrate that generates a red fluorescent product upon interaction with the enzyme.
Formel:
C₁₈H₁₇NO₈ Molecular Weight: 375,333 g/mol |
MDL:
MFCD00064055 CAS-Nummer: 95079-19-9 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...