Resorufin alpha-D-galactopyranoside

Lieferant: AAT BIOQUEST
Achtung

14021
AATB14021EA 460 EUR
AATB14021
Resorufin alpha-D-galactopyranoside
Resorufin alpha-D-galactopyranoside

Resorufin alpha-D-galactopyranoside is a sensitive fluorogenic substrate that generates a red fluorescent product (resorufin) upon interaction with alpha-galactosidase.


  • Alpha-galactosidase fluorogenic substrate
  • Sensitive detection
  • Multiple applications


Bitte beachten For research use only.

Molecular Weight: 375,33 g/mol
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR