FCB [Fluorescein di-beta-D-cellobioside]
Lieferant: AAT BIOQUEST
|
Achtung
|
14025
AATB14025EA
460
EUR
AATB14025
FCB [Fluorescein di-beta-D-cellobioside]
FCB [Fluorescein di-beta-D-cellobioside]
This non-fluorescent fluorescein substrate generates the bright fluorescein product that has Ex/Em = 492/514 nm, and can be easily detected with a FITC filter set.
- Cellulase fluorogenic substrate
- Sensitive detection
- Multiple applications
Bitte beachten For research use only.
Molecular Weight:
980,87 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...