Xite™ Green beta-D-galactopyranoside

Lieferant: AAT BIOQUEST
Achtung

14030
AATB14030EA 292 EUR
AATB14030
Xite™ Green beta-D-galactopyranoside
Xite™ Green beta-D-galactopyranoside

Xite™ Green beta-D-galactopyranoside is a fluorogenic substrate for beta-galactosidase (β-gal).


  • Beta-galactosidase fluorogenic substrate
  • Sensitive detection
  • Multiple applications


Bitte beachten For research use only.

Molecular Weight: 494,5 g/mol
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR