Xite™ Red beta-D-galactopyranoside
Lieferant: AAT Bioquest
14035
AATB14035EA 292 EUR
AATB14035
Xite™ Red beta-D-galactopyranoside
Xite™ Red beta-D-galactopyranoside
Xite™ Red beta-D-galactopyranoside provides a simple and sensitive tool to detect beta-galactosidase (β-gal) activity.
  • Beta-galactosidase fluorogenic substrate
  • Sensitive detection
  • Multiple applications
Bitte beachten For research use only.