Xite™ Red beta-D-galactopyranoside
Lieferant: AAT Bioquest
14035
AATB14035EA
292
EUR
AATB14035
Xite™ Red beta-D-galactopyranoside
Xite™ Red beta-D-galactopyranoside
Xite™ Red beta-D-galactopyranoside provides a simple and sensitive tool to detect beta-galactosidase (β-gal) activity.
- Beta-galactosidase fluorogenic substrate
- Sensitive detection
- Multiple applications
Bitte beachten For research use only.