Cyanine 3-NHS ester chloride ≥90% [equivalent to Cy3® NHS ester]
Lieferant: AAT BIOQUEST
Synonyme:
Cy3 NHS ester, 1-[6-(2,5-Dioxopyrrolidin-1-yloxy)-6-oxohexyl]-3,3-dimethyl-2-[(1E,3E)-3-(1,3,3-trimethylindolin-2-ylidene)prop-1-enyl]-3H-indolium chloride, Cyanine 3 N-hydroxysuccinimide ester, Cyanine 3 monosuccinimidyl ester, eq Cy3® NHS ester
141
AATB141EA
152
EUR
AATB141
Cyanine 3-NHS ester chloride ≥90% [equivalent to Cy3® NHS ester]
Cyanine 3-NHS ester chloride
A variety of cyanine 3 (Cy3®) dyes has been used to label biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis.
- Multiple Bioconjugation Applications
- Excellent Quality
Bitte beachten For research use only.
Formel:
C₃₄H₄₀ClN₃O₄ Molecular Weight: 590,15 g/mol |
CAS-Nummer:
146368-16-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...