Cyanine 3 maleimide ≥90% [equivalent to Cy3® maleimide]
Lieferant: AAT BIOQUEST
Synonyme: Cy3 maleimide
142
AATB142EA 307 EUR
AATB142
Cyanine 3 maleimide ≥90% [equivalent to Cy3® maleimide]
Cyanine 3 maleimide
A variety of cyanine 3 (Cy3®) dyes has been used to label biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis.
  • Multiple Bioconjugation Applications
  • Excellent Quality
Bitte beachten For research use only.