iFluor® 605 succinimidyl ester
Lieferant: AAT BIOQUEST
1420
AATB1420EA 309 EUR
AATB1420
iFluor® 605 succinimidyl ester
iFluor® 605 succinimidyl ester
iFluor® 605 succinimidyl ester is the most convenient reactive form of iFluor® 605 dyes that can be readily used for labeling proteins such as antibodies.
  • Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only
Neuer Artikel bei VWR!