iFluor® 605 maleimide
Lieferant: AAT BIOQUEST
1421
AATB1421EA 309 EUR
AATB1421
iFluor® 605 maleimide
iFluor® 605 maleimide
iFluor® 605 maleimide selectively reacts with thiol group of a biomolecule.
  • Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only