iFluor® 625 maleimide
Lieferant: AAT BIOQUEST
1423
AATB1423EA
309
EUR
AATB1423
iFluor® 625 maleimide
iFluor® 625 maleimide
iFluor® 625 maleimide selectively reacts with thiol group of a biomolecule.
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only