iFluor® 510 succinimidyl ester
Lieferant: AAT BIOQUEST
1424
AATB1424EA
309
EUR
AATB1424
iFluor® 510 succinimidyl ester
iFluor® 510 succinimidyl ester
iFluor® 510 succinimidyl ester is the most convenient form of activated iFluor® 510 derivatives that can be used for labeling proteins such as antibodies.
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only