iFluor® 540 succinimidyl ester
Lieferant: AAT BIOQUEST
1426
AATB1426EA
309
EUR
AATB1426
iFluor® 540 succinimidyl ester
iFluor® 540 succinimidyl ester
iFluor® 540 succinimidyl ester is the most convenient reactive form of iFluor® 540 dyes that can be readily used for labeling proteins such as antibodies.
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only