iFluor® 540 maleimide
Lieferant: AAT BIOQUEST
1427
AATB1427EA 309 EUR
AATB1427
iFluor® 540 maleimide
iFluor® 540 maleimide
iFluor® 540 maleimide selectively reacts with thiol group of a biomolecule.
  • Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only