Eu DBCO, trFluor™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

1446
AATB1446EA 439 EUR
AATB1446
Eu DBCO, trFluor™ fluoreszierende Probe
Eu DBCO

Many biological compounds present in cells, serum or other biological fluids are naturally fluorescent, and thus the use of conventional, prompt fluorophores leads to serious limitations in assay sensitivity due to the high background caused by the autofluorescence of the biological molecules to be assayed.


  • No crosslinking
  • High stability and selectivity
  • Insensitive to fluorescence quenching
  • High emission yield

Molecular Weight: 1765,72 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=88c4afd4a28de989e518ee0e4831afa08c09210e&vid=a48d04e854fe21861c64a3d9ae7fe63485ccbd75&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR