Cyanine 3 amine ≥95% [equivalent to Cy3® amine]
Lieferant: AAT BIOQUEST
Synonyme: Cy3 amine
145
AATB145EA 307 EUR
AATB145
Cyanine 3 amine ≥95% [equivalent to Cy3® amine]
Cyanine 3 amine
A variety of cyanine 3 (Cy3®) dyes has been used to label biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis.
  • Multiple Bioconjugation Applications
  • Excellent Quality
Bitte beachten For research use only.