Cyanine 5 monoacid ≥95% [equivalent to Cy5® acid]
Lieferant: AAT BIOQUEST
Synonyme:
Cy5 acid, 2-(5-(1-(5-Carboxypentyl)-3,3-dimethyl-5-sulphoindolin-2-ylidene)penta-1,3-dien-1-yl)-1-ethyl-3,3-dimethyl-3H-indol-1-ium-5-sulphonate
150
AATB150EA
307
EUR
AATB150
Cyanine 5 monoacid ≥95% [equivalent to Cy5® acid]
Cyanine 5 monoacid
A variety of cyanine 5 (Cy5®) dyes has been used to label biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis.
- Multiple Bioconjugation Applications
- Excellent Quality
Bitte beachten For research use only.
Formel:
C₃₃H₄₀N₂O₈S₂ Molecular Weight: 656,809 g/mol |
MDL:
MFCD29477740 CAS-Nummer: 146368-11-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...