3-Cyan-7-ethoxycumarin ≥95%

Lieferant: AAT BIOQUEST
Achtung

15001
AATB15001EA 120 EUR
AATB15001
3-Cyan-7-ethoxycumarin ≥95%
3-Cyan-7-ethoxycumarin

This fluorogenic coumarin ether is widely used for monitoring P450 activities in cell extracts and solutions.

Formel: C₁₂H₉NO₃
Molecular Weight: 215,21 g/mol
MDL: MFCD00467359
CAS-Nummer: 117620-77-6
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR