Resorufin benzyl ether ≥90%

Lieferant: AAT BIOQUEST
Achtung

15020
AATB15020EA 120 EUR
AATB15020
Resorufin benzyl ether ≥90%
Resorufin benzyl ether

This fluorogenic resorufin ether is widely used for monitoring P450 activities in cell extracts and solutions.

Formel: C₁₉H₁₃NO₃
Molecular Weight: 303,31 g/mol
MDL: MFCD00171613
CAS-Nummer: 87687-02-3
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR