7-Ethoxyresorufin ≥95%
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
Ethoxyresorufin, 7-Ethoxy-3H-phenoxazin-3-one, Resorufin ethyl ether
15023
AATB15023EA
120
EUR
AATB15023
7-Ethoxyresorufin ≥95%
7-Ethoxyresorufin
This fluorogenic resorufin ether is widely used for monitoring P450 activities in cell extracts and solutions.
Formel:
C₁₄H₁₁NO₃ Molecular Weight: 241,246 g/mol |
MDL:
MFCD00037661 CAS-Nummer: 5725-91-7 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...