Resorufin-7-O-methyl ether ≥90%

Lieferant: AAT BIOQUEST
Achtung

Synonyme: Methoxyresorufin

15024
AATB15024EA 120 EUR
AATB15024
Resorufin-7-O-methyl ether ≥90%
Resorufin-7-O-methyl ether

This fluorogenic resorufin ether is widely used for monitoring P450 activities in cell extracts and solutions.

Formel: C₁₃H₉NO₃
Molecular Weight: 227,219 g/mol
CAS-Nummer: 5725-89-3
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR