Pentoxyresorufin ≥95%
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
Resorufin pentyl ether
15025
AATB15025EA
120
EUR
AATB15025
Pentoxyresorufin ≥95%
Pentoxyresorufin
This fluorogenic resorufin ether is widely used for monitoring P450 activities in cell extracts and solutions.
Formel:
C₁₇H₁₇NO₃ Molecular Weight: 283,33 g/mol |
CAS-Nummer:
87687-03-4 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...